Stampa…
Cuvettes for fluorescence
measurements
Fornitore: VARIAN
6610021200 6610021300 6610021400 6610021500 6610021600 6610023500 6610023700 6610024200 6610000900 6610001200
VARI6610021200EA 744 EUR
VARI6610021200 VARI6610021300 VARI6610021400 VARI6610021500 VARI6610021600 VARI6610023500 VARI6610023700 VARI6610024200 VARI6610000900 VARI6610001200
Cuvettes for fluorescence
measurements
Cuvette Cuvette per spettrofotometro
These cuvettes are designed for use with Cary Eclipse fluorescence spectrophotometer.
Applications include academia, Life sciences, biotechnology, chemicals, energy and food analyses.