Stampa…
Masterflex® Sanitary Clamp, Tri-Clamp, 3”
Fornitore: Avantor Fluid Handling
VWR Item#MFLX31201-99
Your Organization's Price:
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance
  • Rated to 150 psi
  • Sterilizable via Autoclave and Gamma Irradiation
  • Features anti-tamper zip-tie lock out
  • Manufactured and packaged in cleanroom
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.