Stampa…
Abaque Peristaltic Hose Pump; 2.42 GPM; 73 RPM; 1 HP 3P
Fornitore: Avantor Fluid Handling
VWR Item#MFLX75810-06
Your Organization's Price:
Pump with confidence with a standard configured system or a custom one.
  • High performance: Seal-less design eliminates leaks and product contamination, and have the ability to run dry continuously without adversely affecting the pump’s performance, pressure, and accuracy (no slip).
  • Reliability: The Abaque pump has none of the usual parts that create problems on other pumps: seats, valves, mechanical seals. The hose is the only replacement part.
  • Handling of the toughest pumping challenges, including extremely abrasive and aggressive fluids, solid loaded liquids, as well as shear sensitive and viscous materials
  • Maximum Discharge Pressure: Models 75810-01 through 75810-06 - 109 PSI(7.5 bar); All other models - 232 PSI(16 bar)
Pumping action is achieved by compression of a circular loop of elastomeric hose with two diametrically opposed rotating shoes. With this rotational motion fluid is pushed ahead inside a hose which is compressed and relaxed by shoes mounted on a rotor. When each shoes reaches the end of the loop, the reinforced tube immediately returns to its original shape, thereby ensuring suction and priming, creating a strong vacuum pulling more fluid inside.<br /><br />Applications<br />• Ceramic: ceramic slips, mold filling, filter press<br />• Mining: sludge, clay slurry, lead sulfate, pyrite slurry<br />• Water Treatment: lime liquor, flocculate dispersion, sludge and slurries<br />• Building Industry: liquid or fibrous mortar, liquid plaster, light concrete<br />• Chemical Industry: acid, alcohol, detergent pumping<br />• Paint & Coatings: water base and acrylic paint, pigments, wall coating<br />• Filter Press: filling and pressing<br />• Paper Industry: latex, slurries, kaolin<br />• Food Industry: tomato sauce, mashed potatoes, gelatin, beer slurries, fish paste<br />• Balneology: thermal muds<br />• Others: glue, liquid salt, seed fertilizer<br /><br />Available options<br />• Leak detector<br />• Vacuum kit<br />• Non-metallic inserts (PPH, PVDF)<br />• Pulsation dampener<br /><br />Hose Materials<br />• Natural rubber: Highly resilient with excellent abrasion resistance and strength.<br />• Buna (NBR): Highly wear resistant to oily products (FDA approved).<br />• EPDM: High chemical resistance, especially with concentrated acids, alcohols, and ketones.<br />• CSM: Resistant to chemicals. High resistance to oxidants, as well as to concentrated bases and acids.
<b>Informazioni per l'ordine: </b><p>Pumps listed are configured for Natural Rubber Hose, and use 316SS Inserts and 3-Phase motor. Other configurations are available to meet specific needs - please call us to review your application.</p>