Stampa…
Masterflex® Variable-Area Flowmeter, Correlated Reading, High-Flow, Brass Housing and Fittings, 65-mm Scale, 42.094 LPM Air
Fornitore: Avantor Fluid Handling
VWR Item#MFLX03268-88
Your Organization's Price:
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.