Stampa…
GoatThroat 2M-BPR 63mm (2.5in) Buttress Adapter With O-ring
Fornitore: Avantor Fluid Handling
VWR Item#MFLX75650-92
Your Organization's Price:
Enhance versitility of your GoatThroat™ Spillproof Barrel Pump.
  • Optimize performance of your GoatThroat™ Spillproof Barrel Pumps
Accessories for GoatThroat Spillproof Barrel Pump optimize performance or suitable for maintenance.<br /><br />The pneumatic air adapter, model 75650-40, allows GoatThroat™ pumps to be used with shop air, air compressors, or bottle compressed air. Uses up to 4 psi to deliver fluids up to 4.5 GPM. External pressure valve keeps containers from being over pressurized.