Stampa…
AW-Lake VCB Sight Flow Transmitter, 0-5 VDC Output, 1-10 GPM, 12-35 VDC
Fornitore: Avantor Fluid Handling
VWR Item#MFLX32825-20
Your Organization's Price:
Output signal for local and remote indication.
  • Easily view flow through plastic lens
These sight flow sensors are designed for low-cost applications where visual indication and sensing output signals are required—available with 4 to 20 mA, pulse, relay, or 0 to 5 VDC output. Durable polypropylene body is compatible with a variety of fluids. Sensors offer a flow measuring accuracy of ±2% full-scale, and are capable of handling pressures up to 150 psig and temperatures up to 150 °F (66 °C).