Masterflex® Ismatec® Quick-Couple Pump Head for 1.6 mm Wall Tubing, 3 Rollers, 3400 mL/min
Fornitore: Avantor Fluid Handling
Snap your pump head onto your drives quickly with turn-and-lock mounting system.
- Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
- 3 rollers
Ensure optimum performance from your Ismatec® pumps.