Stampa…
Masterflex® I/P® Easy-Load® Main Body Assembly, Occlusion Bed and Rear Bearings, PPS Housing, SS Rotor
Fornitore: Avantor Fluid Handling
VWR Item#MFLX77601-67
Your Organization's Price:
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
  • Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.