Stampa…
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-N, L/S 36; 50 ft
Fornitore: Avantor Fluid Handling
VWR Item#MFLX06404-36
Your Organization's Price:
Well-suited for vacuum/pressure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 36
<B>Applications</B>: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance<br /><B>Characteristics</B>: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.