Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-N, L/S 24; 50 ft
Fornitore: Avantor Fluid Handling
Well-suited for vacuum/pressure applications.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 24
<B>Applications</B>: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance<br /><B>Characteristics</B>: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.