Stampa…
HPLC columns, Hypersil GOLD™ C8
Fornitore: Thermo Fisher Scientific
25205-014001 25202-152130 25205-011001 25203-154630 25203-104630 25203-103030 25203-102130 25203-052130 25203-012101 25202-102130 25202-052130 25203-014001 25205-152130 25205-254630 25205-254030 25205-154630 25205-054630 25205-052130 25205-102130 25205-104630 25203-013001 25203-011001 25205-013001 25205-012101 25203-153030 25203-152130 25203-054630 25203-053030
554-4492EA 485 EUR
554-4492 555-7655 554-4487 554-4305 555-7651 555-7675 555-7674 554-4304 554-4482 555-7653 554-4303 554-4484 555-7946 554-4306 555-7901 555-7949 555-7926 555-7923 555-7932 555-7935 554-4483 554-4481 554-4490 554-4488 555-7632 555-7631 555-7671 555-7669
HPLC columns, Hypersil GOLD™ C8
Colonne cromatografiche
Recommended for analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention.
  • Lower hydrophobicity allows compounds to elute quicker from the column
  • Similar selectivity to C18 columns but reduced retention
  • Faster separations, excellent peak shape
  • High efficency, outstanding sensitivity