Stampa…
Digital camera, Axiocam MRc5
Fornitore: ZEISS
0000000450354
ZEIS0000000450354EA 0 EUR
ZEIS0000000450354
Digital camera, Axiocam MRc5
Camere
This high resolution compact digital camera for microscopy can work in a stand-alone mode driver software, standard FireWire/IEEE1394 interface including cable and infrared barrier filter BG40.
  • Blue anodized aluminum, cooling fins, ¼" photo thread for tripod mount housing
  • One stage Peltier cooling control signals
  • 12 different capture modes and fast live images
  • Trigger in/out signal for control of external components
  • Fast live image modes for easy orientation on the specimen