Stampa…
TEVISIO, TVD 750/940/DM Magnifier Luminaire
Fornitore: WALDMANN
113714000-0080
WLDM113714000-0080EA 0 EUR
WLDM113714000-0080
TEVISIO, TVD 750/940/DM Magnifier Luminaire
Lenti d'ingrandimento
The TEVISIO magnifier luminaire delivers delight right across the range. Either in terms of fast and problem-free lens changes, the flexible lens ring with bayonet connection, or the perfectly conceived technical lighting concept or the visualiser function that suddenly also makes unseen errors visible.
  • Film button in luminaire head for On/Off
  • Dimming and visualiser function (segment switching)
  • Materials: colourless anodised aluminium with black special-purpose plastic
  • Arm: spring-relieved with 2D or 3D head joint
Informazioni per l'ordine: DeliveryMore Product Information