Stampa…
Overhead stirrer tools
Fornitore: witeg Labortechnik
DH.WOS01025 DH.WOS01016 DH.WOS01026
WITGDH.WOS01025EA 638 EUR
WITGDH.WOS01025 WITGDH.WOS01016 WITGDH.WOS01026
Overhead stirrer tools
Dispositivi agitatori Accessori di agitazione
These high-speed overhead stirrer tools are designed for mixing applications in the laboratory specialised for low- to medium-viscosity materials.
  • Jog-Shuttle controller with touch-button
  • Maximal viscosity 10000/50000 mPas
  • Chrome-plated stainless-steel chuck to prevent corrosion
  • Impeller height adjustable
  • Six light sources and LCD display
These units provide steady and constant stirring speed and designed with powerful DC motor for various stirring applications such as direct-driven or worm-geared.
The direct driven motor ensures high-speed at allMore Product Information