Stampa…
Dishes, evaporating, quartz glass
Fornitore: witeg Labortechnik
8.651.001 8.651.004 8.651.006 8.651.007 8.651.005
WITG8.651.001EA 0 EUR
WITG8.651.001 WITG8.651.004 WITG8.651.006 WITG8.651.007 WITG8.651.005
Dishes, evaporating, quartz glass
Vassoi/Piatti/Contenitori Bacinelle/piastre di evaporazione
Evaporating dishes, made of quartz glass, with spout.
  • Excellent chemical resistance
  • Excellent temperature resistance (up to 1000 °C)
Certificazioni: DIN 12341