Stampa…
Clamps, three pronged
Fornitore: witeg Labortechnik
8.430.000
WITG8.430.000EA 17.3 EUR
WITG8.430.000
Clamps, three pronged
Morsetti Clamp per uso generico
These universal three-pronged clamps are made of laquered aluminium, with nickel plated brass screws.
  • Jaws are covered with a layer of cork to protect clamped items