Stampa…
Culture flasks, Erlenmeyer shape, BIOGEN
Fornitore: witeg Labortechnik
5.560.250 5.560.050
WITG5.560.250EA 260 EUR
WITG5.560.250 WITG5.560.050
Culture flasks, Erlenmeyer shape, BIOGEN
Fiasche Fiasche per colture
Culture flask, made of borosilicate glass, Erlenmeyer shape, with straight neck for metal caps.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes