Stampa…
Measuring Cylinders, Tall Form, Class A
Fornitore: witeg Labortechnik
3.715.000
WITG3.715.000EA 91.3 EUR
WITG3.715.000
Measuring Cylinders, Tall Form, Class A
Cilindri Cilindri di misurazione
Brown Graduated cylinder class A with spout tall form made of borosilicate glass 3.3/DURAN® components with PE stopper.
Certificazioni: According to DIN ISO 4788