Stampa…
Automatic burettes, Pellet pattern, class B
Fornitore: witeg Labortechnik
3.114.506
WITG3.114.506EA 443 EUR
WITG3.114.506
Automatic burettes, Pellet pattern, class B
Burette
Automatic burettes, Pellet pattern, class B, made of amber borosilicate glass, short line graduation.
  • Stopcock, with hollow NS glass key, between burette and reservoir
  • Lateral stopcock, with solid NS glass key
  • Complete with 2000 ml reservoir bottle, made of DURAN® borosilicate glass, without aspirator bulb