Stampa…
Flasks, round bottom, with standard ground joints
Fornitore: witeg Labortechnik
0.600.010 0.603.025 0.603.025.E 0.603.050.E 0.603.100.E 0.603.250.E 0.603.500.E 0.603.001.E 0.603.002.E 0.602.250.E 0.602.500.E 0.602.001.E 0.602.002.E 0.604.250.E 0.604.500.E 0.604.001.E 0.600.100.E 0.600.250.E 0.601.025.E 0.601.050.E 0.601.100.E 0.601.250.E 0.602.025.E 0.602.050.E 0.602.100.E 0.600.005.E 0.600.010.E 0.600.025.E 0.600.050.E
201-1371EA 84.2 EUR
201-1371 WITG0.603.025 WITG0.603.025.E WITG0.603.050.E WITG0.603.100.E WITG0.603.250.E WITG0.603.500.E WITG0.603.001.E WITG0.603.002.E WITG0.602.250.E WITG0.602.500.E WITG0.602.001.E WITG0.602.002.E WITG0.604.250.E WITG0.604.500.E WITG0.604.001.E WITG0.600.100.E WITG0.600.250.E WITG0.601.025.E WITG0.601.050.E WITG0.601.100.E WITG0.601.250.E WITG0.602.025.E WITG0.602.050.E WITG0.602.100.E WITG0.600.005.E WITG0.600.010.E WITG0.600.025.E WITG0.600.050.E
Flasks, round bottom, with standard ground joints
Fiasche Palloni a fondo tondo
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.