Stampa…
Vacuum stopcocks, angled, Witaflo
Fornitore: witeg Labortechnik
0.290.203
WITG0.290.203EA 52.8 EUR
WITG0.290.203
Vacuum stopcocks, angled, Witaflo
Rubinetti
High vacuum stopcocks, Witaflo-red are made of borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
With adjustable PTFE-seal, leak rate <10-6, usable between -20 and +200 °C, with overtwist safety device.