Stampa…
Shell vials, WHEATON®
Fornitore: DWK Life Sciences
W225243 225126 225129 225242 W225244 225120 225124
WHEAW225243EA 269 EUR
WHEAW225243 WHEA225126 WHEA225129 WHEA225242 WHEAW225244 WHEA225120 WHEA225124
Shell vials, WHEATON®
Vial
These shell vials are made of borosilicate glass or PP.
PE snap plug caps feature a starburst top for easier needle penetration and are available in five colours.