Stampa…
Screw top vials, WHEATON®
Fornitore: DWK Life Sciences
224795
WHEA224795EA 561 EUR
WHEA224795
Screw top vials, WHEATON®
Vial
These vials made of type I borosilicate glass are designed primarily for use in the Waters WISP® 48-position and other autosamplers.
Informazioni per l'ordine: Caps must be ordered separately.
Confezione: Packaged in shrink- wrapped trays of 100 for convenience and cleanliness.