Stampa…
Ampoule snappers, WHEATON®
Fornitore: DWK Life Sciences
177105
WHEA177105EA 54.8 EUR
WHEA177105
Ampoule snappers, WHEATON®
Ampoule Openers
These snapers for ampoules protect against glass splinters and prevent cross contamination.