Stampa…
Qualitative filter papers, Grade 226, Whatman™
Fornitore: Whatman products (Cytiva)
5226-848 5226-330 5226-200 5226-190
WHAT5226-848EA 435 EUR
WHAT5226-848 WHAT5226-330 WHAT5226-200 WHAT5226-190
Qualitative filter papers, Grade 226, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
A general qualitative filter with creped surface. Widely used in industrial or student laboratories for routine filtration.
  • High filtration speed
  • Retains coarse or gelatinous particulates
  • Unique creped gray surface provides optical contrast for white or dark filtrates
The filter is wet-strengthened with a small amount of chemically stable resin to extend the life of the filter and improve handling
Designed for routine and repetitive applications for which an inexpensive product is acceptable, such as qualitative analysis.
Normal qualitativeMore Product Information