Stampa…
Application specific filter papers, grade 0048, Whatman™
Fornitore: Whatman products (Cytiva)
10348903
WHAT10348903EA 304 EUR
WHAT10348903
Application specific filter papers, grade 0048, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
Filter mat made from a mixture of cellulose and polyester. This mat is used for optically testing baby food (artificial milk) for textile fibers.