Stampa…
Reverse osmosis systems, arium® advance RO
Fornitore: Sartorius
H2O-RO-2-B H2O-RO-2-T H2O-RO-3-B H2O-RO-1-T H2O-RO-1-B H2O-RO-3-T
VIVAH2O-RO-2-BEA 8610 EUR
VIVAH2O-RO-2-B VIVAH2O-RO-2-T VIVAH2O-RO-3-B VIVAH2O-RO-1-T VIVAH2O-RO-1-B VIVAH2O-RO-3-T
Reverse osmosis systems, arium® advance RO
Purificatori d'acqua Sistemi per acqua pura e ultrapura
Directly connected to your potable feed water source, this system delivers pure water (Type III) of the highest quality.
  • Touch-function display and intuitive menu navigation for error-free control, even while wearing gloves
  • iJust software: Enhances the quality of purified water, reduces water usage and calculates the optimal cleaning cycles for the system
  • Produced water is stored in the unique, closed arium® bagtank system (not included), which reliably protects purified water from secondary contamination
  • Innovative Bag technology eliminates costly and