Stampa…
Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Ergo
Fornitore: VISION ENGINEERING
SMR212
VISISMR212EA 4823 EUR
VISISMR212
Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Ergo
Microscopi
Mantis Ergo compact ergonomic stereo microscope. Easy to use with interchangeable lenses. Bench stand with integrated substage illumination. Magnification from 3 to 15× (objectives to be added).
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • Three lens multi-view turret
  • Dynamic 3D lighting
  • White/UV lighting option
Eyepiece-less design. Mantis eyepiece-less technology projects a large high quality 3D image directly to your eye, making it easier to see and more comfortable to view than a binocular stereo microscopes.More Product Information