Stampa…
Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Iota
Fornitore: VISION ENGINEERING
SMI212
VISISMI212EA 2859 EUR
VISISMI212
Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Iota
Microscopi
Mantis Iota compact ergonomic stereo microscope. Easy to use with interchangeable lenses. Bench stand with integrated substage illumination. Magnification from 3 to 8× (objectives to be added).
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • Interchangeable lenses
  • Synchronised LED lighting
Eyepiece-less design. Mantis eyepiece-less technology projects a large high quality 3D image directly to your eye, making it easier to see and more comfortable to view than a binocular stereo microscopes. The clarity of the 3DMore Product Information