Stampa…
BarbLock™ Ultra-Secure Tubing Retainers
Fornitore: Saint Gobain Life Sciences
BL135250 BL135500 BL135252AL BL135375AL BL135500AL BL135501AL
VERNBL135250EA 632 EUR
VERNBL135250 VERNBL135500 VERNBL135252AL VERNBL135375AL VERNBL135500AL VERNBL135501AL
BarbLock™ Ultra-Secure Tubing Retainers
Raccordi e connettori
When it comes to retaining flexible tubing and hose onto barbed fittings, BarbLock™ retainers have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. The retainer uses no adhesives or solvents, is ideal for systems transferring delicate liquids or gases.
  • Replaces inconsistent and leak-prone cable ties and traditional clamps
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Supplies 360° compression over the barb and full dynamic retention