Stampa…
Tygon S3™ B-44-5X IB Tubings
Fornitore: Saint Gobain Life Sciences
AMH00018 AMH00029 AMH00038 AMH00046 AMH00053 AMH00064 AMH00070 AMH00074
VERNAMH00018EA 250 EUR
VERNAMH00018 VERNAMH00029 VERNAMH00038 VERNAMH00046 VERNAMH00053 VERNAMH00064 VERNAMH00070 VERNAMH00074
Tygon S3™ B-44-5X IB Tubings
Tubazioni Tubazione di trasferimento
Tygon S3™ B-44-5X IB tubing from Saint-Gobain is clear and flexible with a non-porous bore. It offers outstanding resistance to harsh alkaline cleaners and is unaffected by commonly used sanitisers. Designed specifically for food and beverage dispensing applications, its added reinforcement withstands up to four times the pressure of non-reinforced tubing.
  • All layers compliant with FDA food contact regulations
  • Handles four times the pressure of non-reinforced tubing
  • Improved flexibility at sub-zero temperatures
  • Smooth, non-porous bore resists particle