Stampa…
Biopharmaceutical tubing C-Flex®, Formulation 374
Fornitore: Saint Gobain Life Sciences
374-250-3 374-375-4 374-500-4 374-125-2 374-750-4 374-750-8 374-1000-6 374-188-2 374-250-2 374-250-4 374-375-3 374-625-4 374-125-2S 374-188-2S 374-250-2S 374-250-3S 374-375-4S 374-500-4S 374-125-2M 374-250-2M 374-250-3M 374-375-4M 374-500-4M 374-031-1
228-1442EA 153 EUR
228-1442 228-1443 228-1445 228-1441 VERN374-750-4 VERN374-750-8 VERN374-1000-6 VERN374-188-2 VERN374-250-2 VERN374-250-4 VERN374-375-3 VERN374-625-4 VERN374-125-2S VERN374-188-2S VERN374-250-2S VERN374-250-3S VERN374-375-4S VERN374-500-4S VERN374-125-2M VERN374-250-2M VERN374-250-3M VERN374-375-4M VERN374-500-4M VERN374-031-1
Biopharmaceutical tubing C-Flex®, Formulation 374
Tubazioni Tubazione di trasferimento
Thermoplastic elastomer, clear.
  • Manufactured under strict GMPs guidelines
  • Non pyrogenic, non cytotoxic, non hemolytic
  • Chemically resistant to concentrated acids and alkalis
  • Significantly less permeable than silicone
  • Low platelet adhesion
  • Low protein binding
  • Ultra-smooth inner bore
  • Lot traceable
C-Flex® is a unique, patented thermoplastic elastomer specifically designed to meet the critical demands of the pharmaceutical, research, biopharmaceutical and diagnostics industries. C-Flex® biopharmaceutical tubing has been used by many of the world’s leadingMore Product Information