Stampa…
Biopharmaceutical tubing C-Flex® , Formulation 082
Fornitore: Saint Gobain Life Sciences
082-750-4 082-125-2 082-188-2 082-750-6 082-750-8 082-1000-6 082-500-4M 082-250-2 082-250-3 082-250-4 082-375-4 082-500-4 082-125-2S 082-188-2S 082-250-3S 082-375-2S
VERN082-750-4EA 134 EUR
VERN082-750-4 VERN082-125-2 VERN082-188-2 VERN082-750-6 VERN082-750-8 VERN082-1000-6 VERN082-500-4M VERN082-250-2 VERN082-250-3 VERN082-250-4 VERN082-375-4 VERN082-500-4 VERN082-125-2S VERN082-188-2S VERN082-250-3S VERN082-375-2S
Biopharmaceutical tubing C-Flex® , Formulation 082
Tubazioni Tubazione di trasferimento
Thermoplastic elastomer, clear.
  • Manufactured under strict GMPs guidelines
  • Non pyrogenic, non cytotoxic, non hemolytic
  • Chemically resistant to concentrated acids and alkalis
  • Significantly less permeable than silicone
  • Low platelet adhesion
  • Low protein binding
  • Ultra-smooth inner bore
  • Lot traceable
C-Flex® is a unique, patented thermoplastic elastomer specifically designed to meet the critical demands of the pharmaceutical, research, biopharmaceutical and diagnostics industries. C-Flex® biopharmaceutical tubing has been used by many of the world’s leadingMore Product Information