Stampa…
PPL Tubes, Bond Elut™
Fornitore: VARIAN
7531901C
VARI7531901CEA 152 EUR
VARI7531901C
PPL Tubes, Bond Elut™
Tubi Provette
Bond Elut PPL (Priority PolLutant) is a modified styrene-divinylbenzene polymer that retains polar analytes, including phenols, for excellent absolute recoveries.
  • Modified styrene-divinylbenzene polymer
  • Large particle size allows fast extraction speeds
  • High surface area and capacity for polar analytes
Bond Elut PPL is a styrene-divinylbenzene (SDVB) polymer that has been modified with a proprietary nonpolar surface. PPL will retain even the most polar classes of analytes, including phenols. The large particle size allows ease of flow for viscous orMore Product Information