Stampa…
Labelling tapes, autoclavable
Fornitore: ULTRATAPE
7160OR050-P3D
UTAP7160OR050-P3DEA 720 EUR
UTAP7160OR050-P3D
Labelling tapes, autoclavable
Nastri
Autoclavable vinyl tape with rubber adhesive.
  • Can be written on with a ball point pen
  • Suitable for medical and pharmaceutical applications