Stampa…

Anti-Wolf-Hirschhorn Syndrome Candidate 2 Protein Chicken Polyclonal Antibody

Fornitore: US Biological
W0485-02
USBIW0485-02EA 474 EUR
USBIW0485-02
Anti-Wolf-Hirschhorn Syndrome Candidate 2 Protein Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Wolf-Hirschhorn Syndrome Candidate 2 Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR