Stampa…

Anti-Sialyl Lewis X Mouse Monoclonal Antibody [clone: 9L648]

Fornitore: US Biological
S1013-51B
USBIS1013-51BEA 243 EUR
USBIS1013-51B
Anti-Sialyl Lewis X Mouse Monoclonal Antibody [clone: 9L648]
Anticorpi
Type: Primary
Antigen: Sialyl Lewis X
Clonality: Monoclonal
Clone: 9L648
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR