Stampa…

Anti-MHC Class 1 Chain-related Gene A, B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F158]

Fornitore: US Biological
M3886-75Y3
USBIM3886-75Y3EA 607 EUR
USBIM3886-75Y3
Anti-MHC Class 1 Chain-related Gene A, B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F158]
Anticorpi
Type: Primary
Antigen: MHC Class 1 Chain-related Gene A, B
Clonality: monoclonal
Clone: 10F158
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR