Stampa…

Anti-Interferon Stimulating Gene 15 Rabbit Monoclonal Antibody [clone: 7H28]

Fornitore: US Biological
I7662-94A
USBII7662-94AEA 620 EUR
USBII7662-94A
Anti-Interferon Stimulating Gene 15 Rabbit Monoclonal Antibody [clone: 7H28]
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:25

Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: monoclonal
Clone: 7H28
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse, Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR