Stampa…

Anti-Calcitonin Gene Related Peptide Mouse Monoclonal Antibody [clone: 2Q2268]

Fornitore: US Biological
C0115-01C-100
USBIC0115-01C-100EA 601 EUR
USBIC0115-01C-100
Anti-Calcitonin Gene Related Peptide Mouse Monoclonal Antibody [clone: 2Q2268]
Anticorpi
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Monoclonal
Clone: 2Q2268
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig, Human,
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR