Stampa…

Anti-B Cell Subset Mouse Monoclonal Antibody (Biotin) [clone: 5K110]

Fornitore: US Biological
B0001-65B
USBIB0001-65BEA 647 EUR
USBIB0001-65B
Anti-B Cell Subset Mouse Monoclonal Antibody (Biotin) [clone: 5K110]
Anticorpi
Type: Primary
Antigen: B Cell Subset
Clonality: monoclonal
Clone: 5K110
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Pig
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR