Stampa…

Anti-TREM1 Rabbit Polyclonal Antibody

Fornitore: US Biological
145319
USBI145319EA 0 EUR
USBI145319
Anti-TREM1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR