Stampa…
Low pressure manifolds and Y connectors, Upchurch Scientific®
Fornitore: UPCHURCH SCIENTIFIC
P-150 P-150-01 P-155 P-190 P-190-01 P-191 P-512 P-512-01 P-513 P-514 P-514-01 P-515-01
554-3094EA 267 EUR
554-3094 554-3095 UPCHP-155 554-3098 554-3099 UPCHP-191 554-3117 554-3118 UPCHP-513 554-3119 554-3120 UPCHP-515-01
Low pressure manifolds and Y connectors, Upchurch Scientific®
Raccordi e connettori
PEEK.
  • Pressure rating up to 1000 psi (69 bar) for manifolds and Y connectors for ¹/₁₆" ext. Ø tubing
  • Pressure rating up to 500 psi (34 bar) for manifolds and Y connectors for ¹/₈" ext. Ø tubing