Stampa…
Screw caps for bottles, Upchurch Scientific®
Fornitore: UPCHURCH SCIENTIFIC
A-610 A-620 A-622 A-626 A-627 A-628 A-629 A-630 A-632
UPCHA-610EA 57.8 EUR
UPCHA-610 UPCHA-620 UPCHA-622 UPCHA-626 UPCHA-627 UPCHA-628 UPCHA-629 UPCHA-630 UPCHA-632
Screw caps for bottles, Upchurch Scientific®
Ghiere Tappi per bottiglie
Bottle caps and plugs, manufactured from Tefzel® ETFE and polypropylene, are easy to use, but still allow many connection options.
These bottle caps fits standard thread GL 45 (1 L) and neck GL 38 (4 L) glass bottles. Each cap has three holes. Two of the holes allow tubing to be pushed straight through into the bottle. The third hole, with a Luer taper, can be used for a number of options.