Stampa…
Heat sealer, Impulse
Fornitore: TUTTNAUER
TSM260M
TUTTTSM260MEA 698 EUR
TUTTTSM260M
Heat sealer, Impulse
Saldatrici Sigillatori di sacchetti
Impulse sealing unit for sealing of medical products. Sterilisation of medical instruments is a critically important activity in medicine and dentistry, as well as in veterinary surgeries, hospitals, clinics and laboratories.
  • Fast and reliable impulse sealers
  • Complies with the strictest standards
Sterilisation is effective only if the sterility of the instruments is preserved from the point of sterilisation until they are used. The sealing process ensures that the instruments remain sealed after sterilisation and maintain perfect sterility duringMore Product Information