Stampa…
TISSUE-TROL™ Control Slides Helicobacter
Fornitore: SIGMA ALDRICH MICROSCOPY
TTR006-25EA
TTR006-25EAEA 254 EUR
TTR006-25EA
TISSUE-TROL™ Control Slides Helicobacter
Vetrini Vetrini di controllo per microscopia
Histology slides with paraffin embedded section of mouse intestine tissue containing Helicobacter pylori. Suitable for use with modified steiner-steiner silver stain procedure No. HT101.
Informazioni per l'ordine: Box contains 24 unstained slides plus one reference slide.