Stampa…
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Fornitore: TOSOH Bioscience
0007165
TOYO0007165EA 2060 EUR
TOYO0007165
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Colonne cromatografiche
TSK-GEL® SP-2SW are silica based stainless steel column packed with spherical silica beads which are surface modified with sulfopropyl strong cation exchange groups. Designed for the separation and analysis of small protein, peptides, nucleosides and other biologically active molecules.