Stampa…
Air flow comboKit 2 with Bluetooth®, Testo 440
Fornitore: TESTO
0563.4407
TEST0563.4407EA 1025 EUR
TEST0563.4407
Air flow comboKit 2 with Bluetooth®, Testo 440
Registratori
Airflow Combokit 2 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Long-term monitoring of indoor air quality
  • Combine the vane probe with Bluetooth with the telescope for ceiling outlets
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probe, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality andMore Product Information