Stampa…
Cole Parmer® Block Heat Inserts for BH-200 and BH-250 Series Block Heaters and CB-200 Series Chiller/Heater
Fornitore: Cole Parmer
36610-42 36610-56 36620-52 36610-52 36610-41 36610-46 36610-50 36610-51 36610-53 36610-57 36620-89 36620-92 36620-71 36620-75 36620-76 36620-77 36620-79 36620-80 36620-81 36620-87 36620-61 36620-63 36620-64 36620-65 36620-66 36620-54 36620-55 36620-51 36610-47 36610-41 36610-64
STUA36610-42EA 227 EUR
STUA36610-42 STUA36610-56 TECN36620-52 STUA36610-52 STUA36610-41 STUA36610-46 STUA36610-50 STUA36610-51 STUA36610-53 STUA36610-57 TECN36620-89 TECN36620-92 TECN36620-71 TECN36620-75 TECN36620-76 TECN36620-77 TECN36620-79 TECN36620-80 TECN36620-81 TECN36620-87 TECN36620-61 TECN36620-63 TECN36620-64 TECN36620-65 TECN36620-66 TECN36620-54 TECN36620-55 TECN36620-51 STUA36610-47 TECN36610-41 TECN36610-64
Cole Parmer® Block Heat Inserts for BH-200 and BH-250 Series Block Heaters and CB-200 Series Chiller/Heater
Riscaldatori a blocchi Blocchi
Interchangeable heating blocks offer excellent thermal conductivity.
  • Versatile - interchangeable heating blocks accommodate various tubes and plates
Interchangeable aluminium heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple sizeMore Product Information