Stampa…

Tetraciclina cloroidrato ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

T2525-25G T2525-100G
TCIAT2525-25GEA 41 EUR
TCIAT2525-25G TCIAT2525-100G
Tetraciclina cloroidrato ≥98.0% (by titrimetric analysis)
Tetracycline hydrochloride
Formula: C₂₂H₂₅ClN₂O₈
Punto di fusione: 214°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8
Merck Index: 14,09196
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f6b42b9aec7c1f860b4d1d1fe3deff05c392011&vid=0f6b42b9aec7c1f860b4d1d1fe3deff05c392011&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -240 to -255 deg (C=0.5, 0.1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR